Anti-vibration pad, Richter
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VIBE-1000
DIVEVIBE-1000EA 116 EUR InStock
DIVEVIBE-1000
Anti-vibration pad, Richter
Oberflächenschutz Oberflächenschutz
Richter anti-vibration pad dampens vibrations generated from lab equipment. It creates a stable surface that will keep equipment in place.
  • Small, lightweight design makes transportation between benches and laboratories easy
  • Pads can be stacked for heavier units or laid side-by-side for use with larger equipment
Ideal for use with vortexes, shakers, rockers and microscopes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge