Labels, wash-off
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WASH-2000
DIVEWASH-2000EA 500 EUR InStock
DIVEWASH-2000
Labels, wash-off
Etiketten
These Wash-Off labels and adhesive completely dissolve in any water temperature in under 30 seconds. They readily accept laser printing for quick and easy labeling.
  • FDA-compliant for indirect food contact
  • Autoclavable, resist exposure to steam at 121 °C
  • Storage at room temperature to moderate freezer temperatures
Fits a variety of bottles, flasks, beakers and other laboratory containers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge