Identification cards, multi-coloured, 96-well and 384-well
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WELL-384H WELL-384V WELL-96H WELL-96V
613-1453EA 137 EUR InStock
613-1453 613-1454 DIVEWELL-96H DIVEWELL-96V
Identification cards, multi-coloured, 96-well and 384-well
Titerplatten-Ständer und Well-Orientierungshilfen
Visual aid for pipetting and identification of samples in microplates. Fits most standard 96-well and 384-well plates.
  • Bright colours make it easy to distinguish between columns and rows
Can be used with the Pipette Guide for convenient viewing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge