Clax DS Desotherm 37B2 Detergent for Chemothermal Disinfection
Lieferant: Diversey
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
101106152
DIVY101106152EA 0.1 EUR InStock
DIVY101106152
Clax DS Desotherm 37B2 Detergent for Chemothermal Disinfection
Reinigungsmittel Waschmittel und Oberflächenreiniger
Clax DS Desotherm is a phosphate free complete mainwash detergent with disinfecting properties, specially formulated for use in commercial and on-premise laundries. The product has been developed for automatic and manual dosing and can be applied at all water hardnesses. Clax Desotherm is suitable for washing all white cotton and polycotton fabrics and colourfast articles.
  • Excellent performance on a wide range of stains and types of soil
  • Good whiteness of linen; prevents greying
  • Effective medium temperature bleaching with minimal fabric damage
  • Effective on proteinaceous soil (e.g. blood and food stains)
  • Prevents corrosion of machine parts
  • Provides disinfection at temperatures of 40 °C and higher
Clax DS Desotherm is an enzymatic phosphate free detergent powder, based on an effective blend of surfactants, water hardness sequestrants, anti greying and anti- corrosion agents. The product also contains a medium temperature bleaching system. Thus the product is excellent in fatty/oily soil removal and also in the removal of particulate soil. It can be used at most common water hardness without losing washing performance. The product has been proven to be effectively disinfecting at temperatures ranging from 40 to 60 °C.
The disinfection efficacy has been proven by performing external EN testing. Clax DS Desotherm is a single shot mainwash detergent which may be used for cleaning fabric from hospitals and other healthcare establishments as well as for cleaning hotel linen and workwear.
Product use instruction - Dosing level is dependent on the washing classification. Please refer to the dosing matrix on the label of the bag for normal washing.
For best results:
use softened water
segregate polyester cotton articles from other articles and wash separately
separate white and coloured goods
sub- sort into light, medium and heavily soiled articles
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge