Actival F4r Alkaline Heavy Duty Cleaner, Low Foam
Lieferant: Diversey
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
101107371
DIVY101107371EA 69.6 EUR InStock
DIVY101107371
Actival F4r Alkaline Heavy Duty Cleaner, Low Foam
Reinigungsmittel Waschmittel und Oberflächenreiniger
Heavy-duty, highly alkaline floor cleaner for removal of animal or vegetable oil and fat. Ideal for restaurants, kitchens, food industry, etc.
  • High alkalinity and good emulsifying power
  • Non fragranced
  • Low foaming, fast acting
  • Versatile application
  • W×D×H for bottle: 135×185×283 mm
Benefits:
Quick and thorough removal of animal/vegetable fat and oil
Non tainting, suitable for kitchen/food prep areas
Ideal for use in scrub
Also suitable for wet mopping and manual scrubbing methods
Suitable to be disposed through oil separators
Manual cleaning:
Dose the product into a bucket filled with water and apply the solution
Allow to act and scrub, then remove the soiled solution
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge