Heavy Duty Non Fuming Acidic Descaler, 20 L, Pascal VA5
Lieferant: Diversey
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5600001
DIVY5600001EA 67.6 EUR InStock
DIVY5600001
Heavy Duty Non Fuming Acidic Descaler, 20 L, Pascal VA5
Reinigungsmittel Waschmittel und Oberflächenreiniger
Pascal is a low foaming, heavy duty, non-fuming nitric acid based descaler for use in a wide range of CIP applications in the food and beverage industry.
  • Pascal is highly effective at removing inorganic scale deposits, including calcium oxalate (beer stone)
  • Pascal is low foaming and suitable for use in CIP applications under conditions of high pressure and turbulence
  • Pascal can also be used for open plant cleaning
  • Pascal is highly economical at in use concentrations
  • Pascal is formulated to be non-fuming
  • Pascal is a conductive liquid detergent and suitable for automatic dosing and control
Benefits:
• Highly effective in removing most inorganic scale deposits, improving operational efficiency.
• Can be used for the passivation of new stainless steel CIP and bottle washing installations.
• Reduced amounts of nitrous oxides when handling and using the product, improving operator safety.
• Suitable for automatic dosing and control by conductivity, ensuring consistent delivery of product.
Dimension: 240W×395D×290L mm
Net weight: 26,2 kg
Gross weight: 27,2 kg
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge