Glass and stainless steel cleaner, TASKI Sprint Glass
Lieferant: Diversey
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7513050
DIVY7513050EA 41.5 EUR InStock
DIVY7513050
Glass and stainless steel cleaner, TASKI Sprint Glass
Reinigungsmittel Waschmittel und Oberflächenreiniger
Ammonia-free, glass cleaner to give a non streaking finish on all surfaces including chrome, stainless steel and laminate counter tops.
  • Quick drying
  • Pleasant fragrance with ONT (Odour Neutralising Technology)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge