Lieferant: DELTA T
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A090007
DLTTA090007EA 129 EUR InStock
DLTTA090007
Kühlakkus
37 C Accus maintain a temperature between 36 to 38 °C.
  • High Efficiency
  • Multi-use and disposable solution
  • Secured and safe
  • Control system
  • High availability
  • GDP, FDA, DQ, IQ, PQ, OQ compliance
Closure: aluminum composite foil and protective cap made of plastic
Color: PE natural
Fluid: red
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge