AVM® - Aseptic Validation Medium (Linden Grain Medium), plant-based
Lieferant: DOEHLER GROUP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
850872872
DOEH850872872EA 671 EUR InStock
DOEH850872872
AVM® - Aseptic Validation Medium (Linden Grain Medium), plant-based
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
Powder for the production of vegan AVM® broth to determine the microbiological load as part of a filling line validation under standardised conditions. AVM® is the Döhler trademark for Linden Grain Medium.
  • Without animal-derived ingredients
  • Ready-to-use powder for easy handling
  • Flexible pH adjustment
  • Easy macroscopic evaluation after 5-7 days
This medium allows standardised detection of beverage spoiling bacteria, yeasts and moulds. Desired pH can be adjusted by the customer to achieve high selectivity of beverage spoiling microorganisms.
For validation, the desired quantity of AVM® broth medium passes through the entire filling system and is finally filled into the original container for incubation. After incubation, each individual container is examined macroscopically for turbidity (positive result). This allows a reliable detection of trace and stray infections.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge