DSDM® Dehydrated Culture Media
Lieferant: DOEHLER GROUP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
971231244
DOEH971231244EA 174 EUR InStock
DOEH971231244
DSDM® Dehydrated Culture Media
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Ready-to-prepare dehydrated media for the detection and enumeration of diastatic wild yeast population in brewing culture yeast and beer samples.
  • Powder provides highest flexibility
  • Easier interpretation
  • Incorporated colour indicator
  • High selectivity
  • Functionality checked by strains from practice
  • Effortless an-/ aerobic incubation
DSDM® convinces with highest selectivity and specificity, also for detection in culture yeast. It is mainly used as a liquid medium, but also by adding agar-agar as a solid medium. By selecting appropriate components, only the growth of super-attenuating yeast is promoted, while culture yeast is inhibited. An additional colour indicator helps to interpret the results by showing a colour change.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge