Silikonelastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Lieferant: DOW CORNING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
42417.2
DOWC42417.2EA 0 EUR InStock
DOWC42417.2
Silikonelastomer, Dow Corning® 3-4241 Dielectric Tough Gel
Silikonelastomer
Dow Corning® 3-4241 Dielectric Tough Gel Clear-Green is a two component, heat curing, silicone gel that is used for electrical insulation and protecting electrical components from contaminants, moisture, and stress.
  • Fast heat cure
  • Two parts are blue and yellow and turn green when mixed (1:1)
  • Conditional primerless adhesion at room temperature
  • Mechanical strength
  • UL 94 V-1 Flammability Rating
  • UV indicator for inspection
  • Long working time
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge