Alkohol-Tester, Alcotest® 6820
Lieferant: DRAEGER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8322660
DRAE8322660EA 1307 EUR InStock
DRAE8322660
Alkohol-Tester, Alcotest® 6820
Elektronische Alkoholmessgeräte
Alcotest® 6820 handheld breathalyser specifically for the needs of law enforcement officers to keep the roads safe and free of impaired drivers.
  • Fast, accurate, and reliable results
  • Hygienic and easy to hold and operate with only one hand
  • Resistant to rough weather conditions such as cold or moisture
  • Slide'n'click" mouthpiece and easily snapped into the holder even when it’s dark
The expanded memory capacity is able to record up to 5000 test results. An optical interface enables wireless transmission of data to the mobile printer directly on site.
A LED display and acoustic signals support the visual display and report the completion of a test measurement. Even with a high alcohol content, the sensor provides fast accurate results for both active as well as passive measurements.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge