Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.
For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.
Bestellinformation: Delivery includes 100 pipettes and one bulb dispenser.