Gas cylinder storage boxes
Lieferant: DUPERTHAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7.90.100
DUEH7.90.100EA 716 EUR InStock
DUEH7.90.100
Gas cylinder storage boxes
Möbel Lagerschränke
Storage box is used for storing gas cylinders outdoors in accordance with the regulations.
  • Galvanised sheet steel
  • Cylinder lock as standard fitting
  • Disassembled upon delivery – easy to assemble
Zertifizierungen: Conforms with TRGS 510.
Lieferumfang: Supplied with intermediate shelf.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge