Evaluation Software for Medical Application
Lieferant: EBRO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1340-2460
EBRO1340-2460EA 1510 EUR InStock
EBRO1340-2460
Evaluation Software for Medical Application
Software Labor-Software
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For routine checks.
  • User friendly
  • High-precision measurements
  • Automatic report generation
  • Automatic user-defined calculations
  • Automatic recognition of process cycles
  • Current version: 3.71
Supports: EBI 10, EBI 12, EBI 11, EBI 16, EBI 20, EBI 40, EBI 100, EBI 300, EBI 310. Creation of user-defined templates for specific devices and thermal processes. Three-dimensional display of sensor placement. FDA 21 CFR Part 11 functionality also possible.
Hardware requirements: Processor speed minimum 1,6 GHz, Working memory 2 GB (32 bit), Working memory 4 GB (64 bit) 4 GB free hard disc space, USB A (universal serial bus).
Software requirements: Windows vista 8 (32 bit or 64 bit), windows 10 (32 bit or 64 bit).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge