Robust and Compact High Temperature Data Logger, EBI 11-T210
Lieferant: EBRO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1340-6260
EBRO1340-6260EA 1280 EUR InStock
EBRO1340-6260
Robust and Compact High Temperature Data Logger, EBI 11-T210
Datenlogger
The EBI 11-T210 temperature data logger is ideal for versatile routine checks in the food and pharmaceutical industries. With an extended measuring range (–30 to +140 °C), high–quality PT1000 sensor, very small size that allows it to fit easily into most bottles or small packaging, robust IP 68 protection class, and the option of use in autoclaves, it ensures reliable pasteurisation control and food safety.
  • Rigid, external temperature sensor: Ø 3 mm, 20 mm long
  • Waterproof and vapour-tight
  • Compact size
This data logger was developed for routine checks in the food and pharmaceutical industries, with a special focus on the requirements of canning factories. It enables precise pasteurisation control in cans, jars, and bottles. The robust IP 68 housing ensures reliability and durability. In addition, it is autoclave-proof, which further emphasises its adaptability and reliability. Its very small size allows it to be placed inside small containers or between tightly packed items, ensuring accurate temperature monitoring even in the most space-constrained environments.
Lieferumfang: 1× EBI 11-T210.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
EGÄNZENDE PRODUKTE
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge