Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Lieferant: EDQM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Y0000049
EDQMY0000049EA 245 EUR InStock
EDQMY0000049
Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Standards Organische Standards
Filled under inert atmosphere protected from humidity.
  • In Monograph 1491
  • Solubility in water: 1 - 10 g/l
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge