MyLab™ Electrophoresis Sampler Kit
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100
EDVO100EA 86.9 EUR InStock
EDVO100
MyLab™ Electrophoresis Sampler Kit
Elektrophorese-Systeme Horizontale Gelelektrophoresesysteme
In this experiment kit, students will explore four unique experiments that utilise agarose gel electrophoresis to separate DNA samples. The DNA banding patterns will be analysed within scenarios that examine forensic, medical, and scientific investigations.
These kits can be stored at room temperature. Storage of Ready-to-Load QuickStrip™ samples in the refrigerator is recommended.
Required: DNA electrophoresis, micropipettes: 5 to 50 µl (Optional), white light box, blue light transilluminator (Optional) and microwave or hot plate.
Time required: Complete in 45 minutes
Lieferumfang: Kit includes: Instructions, Ready-to-Load QuickStrip™ DNA samples, UltraSpec-Agarose™, electrophoresis buffer (50X), practice gel loading solution, FlashBlue™ DNA stain and SYBR™ safe stain.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge