Experimentation kits, ion exchange chromatography
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
243
EDVO243EA 149 EUR InStock
EDVO243
Experimentation kits, ion exchange chromatography
Lehrmittel Chemie-Lehrmittel Chromatographie-Lehrmittel
Most molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically. In this experiment, a mixture of 2 chemicals is absorbed onto a solid support ion-exchange column and separated during elution under conditions that influence their net charge.
  • For 6 lab groups
  • Requires 60-90 minutes
Also required: Spectrophotometer and cuvettes, ring stands and clamps, and 5 ml pipettes.
Lieferumfang: Kit includes instructions, ion exchanger, chemical mixture, potassium acetate buffer, and chromatography columns.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge