Experimentation kits, diversity of fish proteins
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
253
EDVO253EA 86.9 EUR InStock
EDVO253
Experimentation kits, diversity of fish proteins
Lehrmittel Biologie-Lehrmittel Genetics Learning Activities
In this experiment, students study the diversity of fish using pre-stained, lyophilised proteins. Total protein from perch, walleye and salmon is extracted and pre-stained using an indicator dye. Each fish protein sample has a characteristic banding pattern when separated by denaturing SDS-polyacrylamide gel electrophoresis, which can be used to identify the specific species.
  • For 6 lab groups (sharing 3 gels)
  • Electrophoresis 60 minutes, staining 20 minutes, destaining 2 hours
Also required: 3 polyacrylamide gels (12%), vertical gel electrophoresis apparatus, power supply, microcentrifuge, white light box, 5 to 50 µl adjustable or 20 µl fixed volume micropipette, fine tips, methanol, and glacial acetic acid.
Lieferumfang: Kit includes instructions, fish LyphoProtein samples, protein molecular weight standards, practice gel loading solution, buffer, and Protein InstaStain.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge