Experimentation kits, PCR-based VNTR human DNA typing
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
334
EDVO334EA 132 EUR InStock
EDVO334
Experimentation kits, PCR-based VNTR human DNA typing
Lehrmittel Biologie-Lehrmittel Lehrmittel zur PCR
In DNA fingerprinting, variable number tandem repeats (VNTR) are used to identify individuals. Using this kit, students type themselves at the D1S80 locus on chromosome 1. This region contains between 14 and 40 copies of a 16 base pair repeat.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Lieferumfang: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge