Experimentation kits, Water Quality Testing III: Multiplex PCR testing of water contaminants
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
953
EDVO953EA 226 EUR InStock
EDVO953
Experimentation kits, Water Quality Testing III: Multiplex PCR testing of water contaminants
Lehrmittel Biologie-Lehrmittel Lehrmittel zur PCR
Drinking water is routinely tested for contamination. If a screening tests positive, more sophisticated tests are required. One such test uses PCR in multiplex format. In this experiment, students test for the presence of three separate, classroom-safe organisms in a water sample using a single PCR reaction.
  • For 5 lab groups
  • Set up 60 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Automatic micropipettes (5 to 50 µl) and tips, waterbath, ice, balance, microcentrifuge, thermal cycler, electrophoresis apparatus, power supply, microwave or hot plate, UV transilluminator.
Lieferumfang: Kit includes instructions, control DNA and primers, DNA ladder, nonpathogenic E. coli, B. subtilis, S. marcenscens, proteinase K, PCR beads, gel loading dye, agarose, electrophoresis buffer, InstaStain® Ethidium Bromide and FlashBlue™ stain.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge