Experimentation kit, bioremediation by oil eating bacteria
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
956
EDVO956EA 142 EUR InStock
EDVO956
Experimentation kit, bioremediation by oil eating bacteria
Lehrmittel Geologie-Lehrmittel Natural Resources Learning Activities
Oil spills cause devastation to the environment, killing sea life, birds, and coastal plants. Spraying areas of contamination with oil-eating microbes accelerates the degradation of the oil. This process is known as bioremediation. In this open-ended experiment, students grow a mixture of oil-eating bacteria and observe their effectiveness at degrading a variety of oils.
  • For 10 lab groups
  • After establishment of cultures, lab requires 50 minutes (can be done over several days or weeks)
Also required: Incubator, growth flasks, vegetable oil (or other food oils).
Lieferumfang: Kit includes instructions, oil-eating bacteria, growth medium, and pipettes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge