Experimentation kits, what is an epidemic and how does an infection spread?
Lieferant: EDVOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S-68
EDVOS-68EA 82.5 EUR InStock
EDVOS-68
Experimentation kits, what is an epidemic and how does an infection spread?
Lehrmittel Biologie-Lehrmittel Physiology and Health Learning Activities
Infectious agents such as bacteria and viruses can spread rapidly through a population and cause widespready disease and death. In this experiment, students use coloured solutions to simulate the spreading of a disease in the classroom.
  • For 10 lab groups
  • Complete in 30 minutes
Lieferumfang: Kit includes instructions, HCl solution, NaOH, colour indicator, test tubes, and pipettes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge