Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
Lieferant: EDWARDS HIGH VACUUM METTLER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H11301019
EDWAH11301019EA 0 EUR InStock
EDWAH11301019
Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
Schmiermittel
Fomblin® perfluoropolyether oil for use in oil sealed rotary vacuum pumps and are practically a direct replacement for mineral oil in terms of viscosity and vapour pressure. Also it contains Drynert fluid which contains anti-rust and anti-wear additives soluble in Fomblin®. These additives cover metallic surfaces with a protective, corrosion resistant film.
It is widely used for semiconductors, chemical pumping, oxygen pumping, radioactive and metallisation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge