Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Chromogenic media for colourful microbial detection, CHROMagar™

Lieferant: CHROMAGAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


ST162 YE492 CP572 AQ056 BC732 CQ382 CQ392 EB042 EC168 EC169-25 EE222 EE223-25 EF322 EF323-25 EF342 EL382 LK970 LM852 MC122 MS25P MS25N PF652 PS832 RR702 RR703-25 SA162 SA163-25 SQ001 TX182 VB912 VB913-25 CP573-25 SU702 SU703-25
ST162EA 654 EUR InStock
ST162 YE492 CP572 AQ056 BC732 CQ382 CQ392 EB042 EC168 EC169-25 EE222 EE223-25 EF322 EF323-25 EF342 EL382 LK970 LM852 MC122 MS25P MS25N PF652 PS832 RR702 RR703-25 SA162 SA163-25 SQ001 TX182 VB912 VB913-25 CP573-25 CHRASU702 CHRASU703-25
Chromogenic media for colourful microbial detection, CHROMagar™
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Chromogenic technology applied to culture media allows you to differentiate and easily identify, inside one single plate, the growth of different microorganisms.

  • Flexibility of use: Prepare only the quantity you need
  • Very easy to prepare
  • Long shelf life: 18 months
  • Cost saving for overall analysis
  • No need for pH adjustment
  • Supplements always included in the pack

Dr A. Rambach, founder of CHROMagar™, developed the use of this technology in food microbiology. The main advantage of using chromogenic media is the ability to distinguish with the naked eye different microorganisms growing on the same plate.

All CHROMagar™ products are dehydrated culture media. The unit size of every packs is indicated in volume (litres or millilitres) that shows the volume of culture medium that you can prepare: With one bottle you can prepare 5 or 25 L of culture media.

After the preparation of the medium starting from the powder, the 5 L pack allows you to prepare around 250 plates, while the 25 L allows you to prepare 1250 plates.

Every chromogen exhibits its own distinctive colour; the result is a very specific and distinctive colouring of different microorganisms which become clearly distinguishable to the naked eye under normal lighting conditions.

The 'Rambach Agar', available in this product range, is still one of the most used and famous chromogenic culture media for detection and isolation of Salmonella spp.

Lieferumfang: Supplements always included in the pack.

Verpackung: 5 L pack = dehydrated culture media to prepare 5 L of agar or broth or 25 L pack = dehydrated culture media to prepare 25 L of agar or broth.


Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge