ESD-Sicherheitsschuhe, Sandalen, mit Klettverschluss, E-Sport, JALAS® 1605
Lieferant: EJENDALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1605-36 1605-37 1605-38 1605-39 1605-40 1605-41 1605-42 1605-43 1605-44 1605-45 1605-46 1605-47
EJEN1605-36EA 83.27 EUR InStock
EJEN1605-36 EJEN1605-37 EJEN1605-38 EJEN1605-39 EJEN1605-40 EJEN1605-41 EJEN1605-42 EJEN1605-43 EJEN1605-44 EJEN1605-45 EJEN1605-46 EJEN1605-47
ESD-Sicherheitsschuhe, Sandalen, mit Klettverschluss, E-Sport, JALAS® 1605
Schuhe und Stiefel
Anti-static low weight ESD safety sandals made from PU-coated and split leather with EVA, polyester based conductive thread insole and polyester-polyamide lining.
  • Shock absorption zone in Ergothan
  • With plastic shank insole and PU outsole
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for indoors, environments with risk for outsole penetration and dry environments.
Colour: Black/grey/red
Zertifizierungen: EN 61340 (ESD); EN ISO 20345:2011; S1 P, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge