ESD-Sicherheitsschuhe, Zenit Easyroll, JALAS® 1738
Lieferant: EJENDALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1738-42 1738-36 1738-37 1738-38 1738-39 1738-40 1738-41 1738-43 1738-44 1738-45 1738-46 1738-47
EJEN1738-42EA 156.98 EUR InStock
EJEN1738-42 EJEN1738-36 EJEN1738-37 EJEN1738-38 EJEN1738-39 EJEN1738-40 EJEN1738-41 EJEN1738-43 EJEN1738-44 EJEN1738-45 EJEN1738-46 EJEN1738-47
ESD-Sicherheitsschuhe, Zenit Easyroll, JALAS® 1738
Schuhe und Stiefel
Anti-static ESD safety shoes made from PU coated split leather with EVA, polyester-based conductive thread insole and merino wool lining.
  • Double shock-absorption zones in Poron® XRD
  • Boa® fastening
  • ProNose toe reinforcement
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for outdoors, indoors and environments with risk for outsole penetration.
Colour: Black/grey/red
Zertifizierungen: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3 SRC
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge