Halbhohe ESD-Sicherheitsschuhe, zum Schnüren, Jupiter, Jalas® 1828
Lieferant: EJENDALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1828-42 1828-44 1828-47 1828-35 1828-36 1828-37 1828-38 1828-39 1828-40 1828-41 1828-43 1828-45 1828-46 1828-48 1828-49 1828-50
EJEN1828-42EA 129.5 EUR InStock
EJEN1828-42 EJEN1828-44 EJEN1828-47 EJEN1828-35 EJEN1828-36 EJEN1828-37 EJEN1828-38 EJEN1828-39 EJEN1828-40 EJEN1828-41 EJEN1828-43 EJEN1828-45 EJEN1828-46 EJEN1828-48 EJEN1828-49 EJEN1828-50
Halbhohe ESD-Sicherheitsschuhe, zum Schnüren, Jupiter, Jalas® 1828
Schuhe und Stiefel
Wide fit, anti-static ESD safety ankle boots made from PU coated leather with EVA, polyester-based conductive thread insole, polyester/polyamide lining plus a heat and oil resistant outsole.
  • Double shock-absorption zones in Poron® XRD®
  • ProNose toe reinforcement
  • Puncture resistant steel midsole
  • Toe protection cap made from aluminium (200 J)
Mainly used for outdoors, environments with risk for outsole penetration and harsh environments.
Colour: Black/grey/yellow
Zertifizierungen: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3, SRC, CI, HRO
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge