Low pH SDS Sample Buffer
Lieferant: EKSIGENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C44807
EKSIC44807EA 197 EUR InStock
EKSIC44807
Low pH SDS Sample Buffer
Puffer Elektrophorese-Puffer Beladungspuffer
SCIEX Low pH SDS Sample Buffer, 140 mL (100 mM Tris-HCl, pH 6.8, 1% SDS) allows you to use SCIEX’s Gold Standard CE-SDS with a low pH SDS sample buffer. Use of this buffer can increase non-reduced purity by decreasing method induced fragmentation. Less variability and lower RSDs means less troubleshooting and faster method development.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge