Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® MP-250 Digital Melting Point Apparatus

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

03010-15 03010-41 03010-51 03010-59
ELET03010-15EA 2400 EUR InStock
ELET03010-15 ELET03010-41 ELET03010-51 ELET03010-59
Cole-Parmer® MP-250 Digital Melting Point Apparatus
Schmelzpunkt-Bestimmungsgeräte
Rotating viewing head ensures each user can analyse samples clearly.

  • View up to three samples simultaneously for greater efficiency
  • Height adjustable arm with viewing magnifier folds away for easy storage
  • Integral light and 8× magnifying lens enhance sample observation
  • Three audible beeps indicate when temperature is stable and ready for sample
  • 4×1 melt memory capacity
  • Adjustable object lens for sharp focus

The MP-250D series digital melting point apparatus allows you to easily view three capillary tube samples simultaneously through the 8× magnifying lens. The adjustable extension arm can be set to different viewing heights making it ideal for multiple users. A rotating viewing head and safety eye piece eliminate glare on the lens and protect you against hot zone.

A 13-key membrane keypad offers easy-to-use menus. The conveniently located push button controls allow temperature recording without looking away from the sample. A digital microprocessor with ±0,1 °C resolution provides fast warm-up and accurate temperature control.

Units can be calibrated by on site or by the manufacturer/service organization. A calibration kit is required if you choose to calibrate on site.

Lens Magnification: 8× (adjustable focus)
Thermometer Range: 1 to 405 °C

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge