Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® SHM-200 Multiposition Extraction Heater/Stirrers

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

03076-13 03076-16 03076-19 03076-22 03076-37 03076-40 03076-43
ELET03076-13EA 4290 EUR InStock
ELET03076-13 ELET03076-16 ELET03076-19 ELET03076-22 ELET03076-37 ELET03076-40 ELET03076-43
Cole-Parmer® SHM-200 Multiposition Extraction Heater/Stirrers
Heizgeräte Heizhauben
Allows liquids with different boiling points to be heated and or stirred next to each other.

  • Stir or heat solutions simultaneously in each position
  • Accommodate flask capacities from 100 to 1000 ml
  • Unique vented airflow ensures the case remains 'cool-to-the-touch'
  • Robust construction permits continuous operation of mantles as required by routine extraction tests
  • Material: Aluminium

Regulating the heat on these multi-position extraction mantles is simple using the built-in solid-state electronic controls. The units accept round bottom flasks ranging from 100 to 1000 ml. Select an extraction heater/stirrer depending on the size of the flask.

Extraction heaters feature an independent heater per mantle, allowing liquids with different boiling points to be heated alongside each other. Each heater has its own control dial and on/off indicator light, and can reach up to a maximum element temperature of 450 °C.

Use the magnetic stirrer to mix sample while heating. Stirring speeds can be set between 50 and 1000 min⁻¹. The three-position models have one stir controller for all three mantles. The six-position models have two stir controllers; one for each set of three mantles.

The lower part of the unit containing the control equipment is safely separated from the upper heating elements by a well-ventilated air space. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Powder-coated aluminum surfaces and polypropylene top cover ensure good chemical resistance and easy cleanup.

Lieferumfang: 3-place units are supplied with 3×0,71 m long rods, 3×magnetic stirring bars, and 1,8 m power cord with EU-plug, the 115 V unit has a US-plug. 6-place units are supplied with 2×0,71 m long rods, 1×1,4 m long rod, support rod clamps, 6×magnetic stirring bars, and 1,8 m power cord with EU-plug.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge