Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® HM-200 Multi-Position Extraction Heaters

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

03076-01 03076-04 03076-07 03076-10 03076-25 03076-31 03076-34
ELET03076-01EA 2260 EUR InStock
ELET03076-01 ELET03076-04 ELET03076-07 ELET03076-10 ELET03076-25 ELET03076-31 ELET03076-34
Cole-Parmer® HM-200 Multi-Position Extraction Heaters
Heizgeräte Heizhauben
Allow liquids with different boiling points to be heated next to each other.

  • Vented airflow ensures the case remains 'cool to the touch'
  • Heat your solutions simultaneously in each position
  • Robust construction permits continuous operation of the mantles as required by routine extraction tests

Regulating the heat on these multi-position extraction mantles is simple using the built-in solid-state electronic controls. The units accept round-bottom flasks ranging from 100 to 1000 ml. Select an extraction heater depending on the size of your flask.

Extraction heaters feature an independent heater per mantle, allowing liquids with different boiling points to be heated alongside each other. Each heater has its own control dial and on/off indicator light, and can reach up to a maximum element temperature of 450 °C (842 °F).

The control panel is safely separated from the heating elements by a well ventilated air space. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Powder coated aluminium surfaces and polypropylene top cover ensure good chemical resistance and easy cleanup.

Lieferumfang: Supplied with two 71 cm long rods, one 140 cm long rod, support rod clamps, and 1,8 m power cord with EU-plug.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge