Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Stirring/Heating Mantles, SHM-200

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

04641-05 04641-15 04641-25 04641-35 04641-45 04641-55
ELET04641-05EA 1410 EUR InStock
ELET04641-05 ELET04641-15 ELET04641-25 ELET04641-35 ELET04641-45 ELET04641-55
Stirring/Heating Mantles, SHM-200
Heizgeräte Heizhauben
Provides powerful stirring speeds up to 520 min⁻¹ and heating up to 450 °C.

  • Bidirectional stirring with magnetic stirring bar auto capture function
  • Thermal insulating cartridge provides maximum heat transfer
  • Chemical resistant polypropylene housing is 'safe to the touch'

Stir samples while heating up to 450 °C (842 °F). These durable stirring/heating mantles feature a bidirectional magnetic stirrer with speeds up to 520 min⁻¹. An 'auto-recapture' function reactivates stirring if it is interrupted or lost for any reason.

A built-in power controller with dial markings from one to ten offer great repeatability. Two coloured lights indicate 'power on' and 'heat on'. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.

Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.

Lieferumfang: Supplied with 1× magnetic stirring bar and a power cord with UK plug.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge