Cole-Parmer® HM-200 Heating Mantles
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36002-10 36002-14 36002-18 36002-02 36002-06 36002-30 36002-22 36002-26
ELET36002-10EA 837 EUR InStock
ELET36002-10 ELET36002-14 ELET36002-18 ELET36002-02 ELET36002-06 ELET36002-30 ELET36002-22 ELET36002-26
Cole-Parmer® HM-200 Heating Mantles
Heizgeräte Heizhauben
Thermal insulating cartridge provides maximum heat transfer.
  • Chemical resistant polypropylene housing is 'safe to the touch'
Quickly and safely heat round-bottom flasks to 450 °C (842 °F). These simple-to-use heating mantles feature a built-in power controller with dial markings from 1 to 10 for repeatability. An amber coloured 'heater on' light and a clear-coloured 'power on' light indicate when the heating mantle is in use. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Ventilation slots and ceramic fibre insulation keep the chemical resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge