Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® MSHM-200 Magnetic Stirring/Heating Mantles

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

36013-00 36013-02 36013-04
ELET36013-00EA 1690 EUR InStock
ELET36013-00 ELET36013-02 ELET36013-04
Cole-Parmer® MSHM-200 Magnetic Stirring/Heating Mantles
Heizgeräte Heizhauben
Magnetic stirrer and heating mantle in one convenient unit.

  • Flexible coiled heating element wraps the heater around round-bottom flask to optimise heat transfer
  • 'Cool to the touch' exterior provides safe handling
  • Material: Aluminium

These stirring/heating mantles combine the functionality of a heating mantle with a magnetic stirrer providing even heating and mixing. Element temperatures can reach up to 450 °C (842 °F ) yet the exterior remains cool to the touch. For maximum safety, the flask does not come in direct contact with the heating element. Built-in energy regulator and safety earth (ground) screen offer added safety protection.

Reach stirring speeds up to 2000 rpm. Simply use the auto-recapture switch to reactivate stirring if stirring is interrupted and stirring motion is lost. Two coloured lights by each control dial indicate 'power on' and 'heat on'. Powder coated aluminium housing makes it easy to clean. All heating mantles feature a support bracket for holding a support rod.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge