Cole-Parmer® HM-200 Large Volume Heating Mantles
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36013-16 36013-18 36013-20 36013-22
ELET36013-16EA 5980 EUR InStock
ELET36013-16 ELET36013-18 ELET36013-20 ELET36013-22
Cole-Parmer® HM-200 Large Volume Heating Mantles
Heizgeräte Heizhauben
Accommodate 60° funnels, pear-shaped flasks and round-bottom flasks.
  • Bottom opening allows use with 60° funnels of various diameters
  • Accept large capacity flasks -between 10 to 22 litres
  • Three separate electrical circuits allow controlled heating of top, middle and bottom of flasks independently
The 'V-shaped' design of the heating mantle accommodates both pear-shaped and round bottom flasks. Coiled heating element, suspended within a thermal insulating material, provides optimal heat transfer and support. The outer casing remains cool to the touch even when the heating element temperature is at 450 °C (842 °F).
Control heat throughout the mantle using three separate electrical circuits each controlled with its own dial switch. This enables independent temperature control of top, middle and bottom of the flask. Two coloured lights by each control dial indicate 'power on' and 'heat on'. Powder coated aluminium housing makes it easy to clean. Heating mantles feature three support rod clamps that can hold 1,27 cm dia support rods.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge