Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® SHM-250 Digi-Mantle Heaters

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

36204-01 36204-05 36204-09 36204-13 36204-15 36204-19 36204-23
ELET36204-01EA 1610 EUR InStock
ELET36204-01 ELET36204-05 ELET36204-09 ELET36204-13 ELET36204-15 ELET36204-19 ELET36204-23
Cole-Parmer® SHM-250 Digi-Mantle Heaters
Heizgeräte Heizhauben
Heating mantles with interchangeable cartridges.

  • Set speed and temperature in seconds with the intuitive display
  • Complete system requires controller and heating cartridge
  • Achieve uniform temperature up to 450 °C across round bottom flask
  • Stirring speeds from 100 to 2000 min⁻¹ with excellent stability
  • Vented casing keeps heating cartridge cool to the touch
  • Modular design allows different size heating cartridges to be used on the same Digi-mantle controller

Control temperature and stirring speed independently on the Digi-Mantle digital controller. The control panel features indicator lights for power, heater and stirrer. Microprocessor controlled heater displays the heat setting as a percentage of the total power. View the actual temperature (up to 220 °C) of the flask content on a 3-digit LCD display by using the included temperature probe.

Lieferumfang: Order controller and cartridges separately or heating kit includes a controller and a heating cartridge that accommodates 100 to 250, 500 or 1000 ml round bottom flask. Due to its modular design a different size heating cartridge can be used with the same controller, saving you money.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge