Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® HT-200 Grounded Heating Tapes

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

36220-10 36220-12 36220-00 36220-04 36220-06 36220-08 36220-02 36220-15 36220-14
ELET36220-10EA 1020 EUR InStock
ELET36220-10 ELET36220-12 ELET36220-00 ELET36220-04 ELET36220-06 ELET36220-08 ELET36220-02 ELET36220-15 ELET36220-14
Cole-Parmer® HT-200 Grounded Heating Tapes
Klebebänder
Flexible heating tapes are designed to fit metal pipes and glassware.

  • Maximum element temperature of 842 °F (450 °C)
  • Braided earth wire beneath the outer sheath
  • Approximately 25 mm wide

The flexible fiberglass heating tapes are ideal for heating columns, pipes, valves, and transfer lines, where applications need to be observed. The heating element and braided ground wire are wound on a fiberglass core and covered in a braided fiberglass outer sleeve. Tapes are not water-resistant; use in dry conditions only. Tapes are supplied with 3,5'' (9 cm) insulated unheated flexible leads at each end.

Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge