Cole-Parmer® Integrity 10 Reaction Station
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36630-00
ELET36630-00EA 79220 EUR InStock
ELET36630-00
Cole-Parmer® Integrity 10 Reaction Station
Rührer Magnetrührer mit Heizplatte
Use the Integrity 10 reaction station to run ten independent experiments at once. By setting the temperature and stir rate individually for each cell users can assess the impact of both at the same time, speeding up investigative chemistry by ten-fold.
  • Perform 10 independent experiments at once, saving both time and money
  • View experiment results in real time on the monitor
  • Easily transfer data to a PC via a USB port or SD card
  • Variable stirring speeds from 350 to 1200 rpm ensure homogeneous sample mixing
  • Set sample temperature between –30 and 150 °C, with ±0,5 °C temperature accuracy
  • Optional attachment available for refluxing and working under inert conditions
  • Three year warranty
  • New, improved version of PS20001
Easily set up test parameters using the user-friendly touchscreen menu. Intuitive icons reduce potential for costly errors while preprogrammed profiles for routine measurements save time. Store results with information such as reaction composition, experiment owner, concentration, and the name of the profile with data stamp. View the progress of the reaction within each cell on the large monitor. It can also display at-a-glance results graphically. Extensive storage capacity allows for data capture and review. Download data to a PC via a USB port or SD card. Order software (ELET36630-92) for more in-depth analysis and evaluation.
Control the temperature for each cell through the block or by using a temperature probe in each cell's solution. A ±0,5 °C temperature stability and a minimal set point overshoot of 0,5 °C ensure temperature accuracy. Set ramps between 0,1 °C/min. to 5 °C/min. for fast cooling and heating. Use the crash function for even faster temperature changes, which is ideal for kinetic determinations. Individually controlled magnetic stirrer can be set for speeds between 350 to 1200 min⁻¹ (rpm) (use 12×4,5 mm cylindrical or 10×6 mm oval stir bars only). Order reducing sleeves and test tubes to accommodate sample volumes down to 1 ml.
The reflux/inerting head (ELET36630-97) is ideal for running parallel reactions for a long time, as is commonly required in organic chemistry experiments. Use with inerting caps to provide inert atmosphere to all ten cells. The integrated design eliminates the need to connect individual gas lines to each cell for a safer and effective solution.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge