Reaction Stations, RS-200 and RS-250
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36630-01 36630-11
ELET36630-01EA 17200 EUR InStock
ELET36630-01 ELET36630-11
Reaction Stations, RS-200 and RS-250
Rührer Magnetrührer mit Heizplatte
Run multiple reactions in a single station, saving time and money.
  • Control and maintain the same temperature and stirring rate at each position
  • Excellent temperature uniformity across the block
  • Simple menus and touchpad interface to control the system performance
  • Exterior remains cool to the touch for added safety
Run these reaction stations at the same temperature and stirring rates at several positions simultaneously. They are great for a wide range of applications from simple synthesis to process optimisation. High-performance magnetic stirrers ensure maximum coupling between the stirring bar in the sample and motor.
The RS-200 series reaction stations feature a modular design that accommodates a variety of cartridges and tubes. All heating blocks, glassware and reflux heads are interchangeable to accommodate sample volumes ranging from 1 to 1000 ml. All heater cartridges feature 'in block heaters' for temperatures up to 220 °C. Use with the included temperature probe for ±0,5 °C (±0.9°F ) temperature accuracy. Connect cooling plug 36630-71 and chiller to the cartridge to achieve temperature down to –30 °C without the need for an ice bath. A central gas port, combined with gas-tight PTFE caps, allows a vacuum to be pulled or an inert gas to be introduced.
The RS-250 series reaction stations feature a compact footprint for easy integration onto a robotic platform. Use the RS-232 or RS-485 connections to interface with a robotic workstation or a computer.
Lieferumfang: The RS-200 Reaction station (10×25 ml) is supplied with reflux inerting head (36630-72), 10× glass vessels (36630-85), 10× inerting caps (36630-74), 10× magnetic stirring bars (36630-98), temperature probe (36630-70), one pack of cut tubing, and power cord with UK plug. The RS-250 Reaction station (10×25 mm) is supplied with 10× magnetic stirring bars and a 2 m power cord with UK plug. The RS-250 Reaction station (6×57,5 mm) unit is supplied with 6× magnetic stirring bars and a 2 m power cord with UK plug.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge