Drucken
Cole-Parmer® FB-200 Tissue Float Bath
Lieferant: Cole-Parmer
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
48572-55
ELET48572-55EA 1530 EUR
ELET48572-55
Cole-Parmer® FB-200 Tissue Float Bath
Bäder Wasserbäder für Gewebeschnitte
Designed to assist in tissue preparation in histology and pathology laboratories.
  • Easily adjust temperature between 30 to 70 °C
  • The wide-rimmed edge serves as drying surface for processed slides
  • Fast heat up time - heat 2,25 L of water to 45 °C within 40 minutes
  • Temperature stability of ±0,5 °C
  • Aluminium base with PTFE coated interior
Use the tissue float bath before the paraffin sections are placed on slides to prevent wrinkles and folds. The black PTFE interior and rim reduce eye strain by cutting down on glare while improving visibility of tissue sections. White powder-coated aluminium exterior is easy to keep clean. Safety features include an integrated indicator light alerting you when unit is on, as well as automatic shut-off.
Bath lid and thermometer to help maintain desired temperature are available to order separately. Order gelatine to increase surface tension of water in flotation bath.
Lieferumfang: Supplied with three-wire cord with plug.