Cole-Parmer® WD-200 Wax Dispenser
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
85230-07
ELET85230-07EA 5580 EUR InStock
ELET85230-07
Cole-Parmer® WD-200 Wax Dispenser
Paraffinwachs-Trimmer und -Spender
Prevent wax blockage with heated tap.
  • Accurate temperature control with digital display
  • Constructed of stainless steel with antimicrobial coating to inhibit bacteria growth
  • Enhanced safety with high temperature cut-off at 120
  • °C
  • Fully insulated tank prevents heat loss and ensures the outer surface stays cool
This paraffin wax dispenser provides ultrafast heating for on-demand molten wax, ideal for your histology and pathology applications. The heated tap prevents blockage providing a smooth even flow of wax every time. The nondrip lever has three settings—off, on-demand pull dispense, and continuous flow. The 0,5 mm mesh filter screen prevents coarse particles from entering the tap.
Temperature can be set from ambient to 70 °C using digital temperature control panel. The on/off temperature control keeps accuracy to ±1 °C. The 7,5 L capacity tank can melt up to 13,2 lbs. (6 kg) of pelletised wax at a time. When set at 65 °C, the unit will take approximately 90 minutes to fully melt 11 lbs. (5 kg) of pelletised wax.
Lieferumfang: Power cord.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge