Water purification systems, PURELAB® Quest
Lieferant: ELGA LabWater
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PQDIUVM1
ELGAPQDIUVM1EA 5339 EUR InStock
ELGAPQDIUVM1
Water purification systems, PURELAB® Quest
Wasseraufbereitungssysteme Reinstwassersysteme
Ideal water system for a new start up lab. Advanced and affordable, providing tap to Type I water.
  • Maximize your budget: one system delivering three water types
  • Compact: wall or bench mount
  • Sustainable design: made with 85% recycled materials
  • Intuitive: simple to use and install
  • Manual and autovolume dispensing with recirculation of the water from the reservoir up to the point of dispense
  • Integrated 7 liter reservoir
  • USB data port/Hubgrade enabled
The PURELAB® Quest offers three water types, multiple quality sensors, clear information, a compact and economical design, ease of use, a fast flow rate, reliability, and simple maintenance.
The system has been tested through 150000 cycles of dispensing, which is equivalent to over 20 years of lab use. Research and development are at the heart of ELGA. Our global network of engineers and scientists are dedicated to lab water applications.
PURELAB® Quest's components have been proven in tens of thousands of water systems globally and result from over a decade of user experience and feedback. While discovery and research can take a lifetime, PURELAB® Quest will deliver the water you need within seconds. Every day is a Quest.
Made from >85% reclaimed materials, which is a better investment for the planet.
Zertifizierungen: UL/CSA listed. CE marked.
Zubehörinformationen: When purchased as a kit, the system comes with a pretreatment filter and a 0,2 µm capsule filter or a BioFilter. For Type II and III water, the addition of a 15 or 30 liter reservoir will be required. Installation labor can be ordered separately. Please contact your Avantor representative for more information.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge