Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Elmasonic EASY Ultrasonic Baths

Lieferant: ELMA SCHMIDBAUER

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

1118881 1118883 1118885 1118887 1117697 1118890 1118892 1118894 1118896 1118898 1118900 1118902
ELMA1118881EA 305 EUR InStock
ELMA1118881 ELMA1118883 ELMA1118885 ELMA1118887 ELMA1117697 ELMA1118890 ELMA1118892 ELMA1118894 ELMA1118896 ELMA1118898 ELMA1118900 ELMA1118902
Elmasonic EASY Ultrasonic Baths
Ultraschallbäder
The Elmasonic EASY ultrasonic bath series includes 9 units of different sizes and is characterised by simple, user-friendly operation. With 100% ultrasonic power at a frequency of 37 kHz, the EASY solves cleaning tasks quickly and efficiently. Ideal for removing tenacious, mineral soiling and polishing pastes.

  • 37 kHz ultrasonic frequency (control range −2,5 / +5,5 kHz)
  • 2 ultrasonic modes: dynamic mode or eco mode
  • Electronic selector switches on wipeable membrane keypad and LED display of all operating states
  • Electronic temperature control adjustable in steps of 5 °C from 30 to 80 °C for devices with heating (temperature tolerance −5 / +8 °C)
  • Safety switch-off with wake-up function after 8 hours of inactivity or in case of a brief interruption of the power supply
  • Ultrasonic operation automatically shuts down at bath temperature >90 °C
  • Stainless steel splash-proof housing and cleaning tray
  • Side stopcock for emptying the cleaning tank via the rear drain (from Elmasonic EASY 60 H)

Elmasonic EASY the powerful all-rounder has 2 ultrasonic modes for need-based sound field distribution in the ultrasonic bath.
*Dynamic mode, automatically set: Serves more intensive cleaning with higher cleaning power.
*Eco mode, can be switched on: Serves gentle cleaning with quieter operation.

Units feature pre-selection of cleaning time: Cleaning time is adjustable between 1 and 30 minutes or continuous operation (ultrasonic continuous operation max. 6 hours).

2-year warranty from the initial operation, but no longer than 3 years from the production date.

Lieferumfang: Supplied with cover.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge