Pneumatically operated diaphragm valves, Integra®
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
202-128-01
ENTE202-128-01EA 0 EUR InStock
ENTE202-128-01
Pneumatically operated diaphragm valves, Integra®
Ventile Mikroventile
Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution systems. Valve design allows ultrapure water to continuously bypass the diaphragm, eliminating water stagnation and bacteria formation in the valve. Manually adjustable, integral needle valve provides precise control of bypass flow.
  • All wetted parts are PFA and PTFE
  • No external metal components
  • Unique top loading design allows for field service
  • Espy® sensor can be added for electronic monitoring of valve position
¹/₂" orifice, 2-way, adjustable bypass, position indication option.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge