Pneumatically operated diaphragm valves, Integra®
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
202-86-01
ENTE202-86-01EA 0 EUR InStock
ENTE202-86-01
Pneumatically operated diaphragm valves, Integra®
Ventile Mikroventile
Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution systems. Three-ported directional valve enables simplification of piping system design while minimising its footprint. All PFA wetted surfaces for ultrapure and corrosion resistant performance.
  • No external metal parts.
  • Unique top loading design provides for easy field service
  • Valve body design allows significant cavity drainage to minimize fluid entrapment
  • Integrally molded Flaretek® flared tube and PureBond® fusible pipe port connection options
  • Adding Espy® sensors allows for electronic monitoring of valve position
¹/₄" orifice, 3-way, position indication option.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge