Wafer carrier handles for 76,2 mm (3") wafer handling
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A82-03-0215
ENTEA82-03-0215EA 0 EUR InStock
ENTEA82-03-0215
Wafer carrier handles for 76,2 mm (3") wafer handling
Wafer-Handling-Zubehör Wafer-Carrier
  • Attaches to the notches on both end walls of carrier
  • Overall size: 165×91×157 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge