Manifold, Impact® ST
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DZUM044C1
ENTEDZUM044C1EA 0 EUR InStock
ENTEDZUM044C1
Manifold, Impact® ST
Verteiler
Stainless steel manifold for use with Impact® Plus disposable filters.
  • Rapid filter changeout with minimal dripping: no tools and no draining required
  • Compact design provides easy installation or retrofit into existing tools
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge