Round process tank
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E14-35-01-0215
ENTEE14-35-01-0215EA 0 EUR InStock
ENTEE14-35-01-0215
Round process tank
Tanks Tanks, zylindrisch
Injection molded round tank, in PFA material.
  • Tanks should be properly supported in heated applications
  • Maximum recommended temperature limit for PFA tanks is 180 °C
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge